Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMNDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102320
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102320 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102320 Supplier Invitrogen™ Supplier No. PA5102320
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SMNDC1.

This gene is a paralog of SMN1 gene, which encodes the survival motor neuron protein, mutations in which are cause of autosomal recessive proximal spinal muscular atrophy. The protein encoded by this gene is a nuclear protein that has been identified as a constituent of the spliceosome complex. This gene is differentially expressed, with abundant levels in skeletal muscle, and may share similar cellular function as the SMN1 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMNDC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SMNDC1
Gene Accession No. O75940, Q4QQU6, Q8BGT7
Gene Alias 2410004J23Rik; 30 kDa splicing factor SMNrp; 4933440I19Rik; MGC106917; MGC112663; SMN related protein; SMNDC1; SMNR; SMN-related protein; Spf30; splicing factor 30, survival of motor neuron-related; survival motor neuron domain containing 1; survival motor neuron domain-containing protein 1; Survival of motor neuron-related-splicing factor 30; TDRD10; TDRD16C; tudor domain containing 16C; wu:fb37h07; wu:fc23a07
Gene Symbols SMNDC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SMNDC1(Accession O75940), corresponding to amino acid residues L45-E95.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10285, 287768, 76479
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.