Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMNDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144105
Description
SMNDC1 Polyclonal Antibody for IHC (P), IP, ELISA
This gene is a paralog of SMN1 gene, which encodes the survival motor neuron protein, mutations in which are cause of autosomal recessive proximal spinal muscular atrophy. The protein encoded by this gene is a nuclear protein that has been identified as a constituent of the spliceosome complex. This gene is differentially expressed, with abundant levels in skeletal muscle, and may share similar cellular function as the SMN1 gene.
Specifications
SMNDC1 | |
Polyclonal | |
Unconjugated | |
SMNDC1 | |
2410004J23Rik; 30 kDa splicing factor SMNrp; 4933440I19Rik; MGC106917; MGC112663; SMN related protein; SMNDC1; SMNR; SMN-related protein; Spf30; splicing factor 30, survival of motor neuron-related; survival motor neuron domain containing 1; survival motor neuron domain-containing protein 1; Survival of motor neuron-related-splicing factor 30; TDRD10; TDRD16C; tudor domain containing 16C; wu:fb37h07; wu:fc23a07 | |
Rabbit | |
Protein G | |
RUO | |
10285 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
O75940 | |
SMNDC1 | |
Recombinant Human Survival of motor neuron-related-splicing factor 30 protein (1-238AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction