Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113312
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113312 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113312 Supplier Invitrogen™ Supplier No. PA5113312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMO Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Smoothened drosophila homolog (SMOH) is a serpentine protein belonging to the G-protein coupled receptor Fz/Smo family with 787 aa, seven transmembrane G protein-coupled receptors with 4 glycosylation sites and a putative extracellular amino terminus. Smoothened activated by b-Arrestin 2 and GRK2 acts as a receptor for Shh, the Sonic hedgehog gene product, and is a positive regulator of hedgehog signaling pathway along with the smoothened inhibitor patched protein (PTCH) and regulates vertebrate developmental processes, cellular proliferation and differentiation by activating transcription factor Gli. Smoothened mutants are known to cause sporadic basal cell carcinoma of the skin. It is known to express widely in heart and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMO
Applications ELISA, Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SMO
Gene Accession No. P56726, Q99835
Gene Alias bent body; bnb; E130215L21Rik; frizzled family member 11; FZD11; Gx; HGNC:11119; Protein Gx; Protein SMOH; seven transmembrane helix receptor; SMO; SMOH; smoothened; Smoothened homolog; smoothened homolog (Drosophila); smoothened, frizzled class receptor; smoothened, frizzled family receptor; smoothened, seven transmembrane spanning receptor
Gene Symbols SMO
Host Species Rabbit
Immunogen Recombinant Human Smoothened homolog protein (28-233AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319757, 6608
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.