Learn More
Invitrogen™ SMO Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534217
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Bat, Hamster, Elephant, Panda, Horse, Rabbit, Pig, Platypus (100%); Opossum (93%); Turkey, Chicken, Xenopus (87%); Zebrafish, Seq squirt (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Smoothened drosophila homolog (SMOH) is a serpentine protein belonging to the G-protein coupled receptor Fz/Smo family with 787 aa, seven transmembrane G protein-coupled receptors with 4 glycosylation sites and a putative extracellular amino terminus. Smoothened activated by b-Arrestin 2 and GRK2 acts as a receptor for Shh, the Sonic hedgehog gene product, and is a positive regulator of hedgehog signaling pathway along with the smoothened inhibitor patched protein (PTCH) and regulates vertebrate developmental processes, cellular proliferation and differentiation by activating transcription factor Gli. Smoothened mutants are known to cause sporadic basal cell carcinoma of the skin. It is known to express widely in heart and brain.
Specifications
SMO | |
Polyclonal | |
Unconjugated | |
SMO | |
bent body; bnb; E130215L21Rik; frizzled family member 11; FZD11; Gx; HGNC:11119; Protein Gx; Protein SMOH; seven transmembrane helix receptor; SMO; SMOH; smoothened; Smoothened homolog; smoothened homolog (Drosophila); smoothened, frizzled class receptor; smoothened, frizzled family receptor; smoothened, seven transmembrane spanning receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100033990, 100352661, 100515110, 100762005, 25273, 319757, 482262, 539308, 6608 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P56726, P97698, Q99835 | |
SMO | |
Synthetic 15 amino acid peptide from N-terminal extracellular domain of human SMO. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Bat, Hamster, Elephant, Panda, Horse, Rabbit, Pig, Platypus (100%); Opossum (93%); Turkey, Chicken, Xenopus (87%); Zebrafish, Seq squirt (80%). | |
50 μg | |
Primary | |
Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Bat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.