Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMOC-1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP215123

Catalog No. NBP215123


Only null left
Add to Cart

Description

Description

SMOC-1 Polyclonal specifically detects SMOC-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SMOC-1
Polyclonal
Unconjugated
PBS, 1% BSA, 20% Glycerol (pH7) with 0.025% Proclin 300
Secreted modular calcium-binding protein 1, SMOC-1, SPARC related modular calcium binding 1, SPARC-related modular calcium-binding protein 1
Rabbit
48 kDa
0.1 mL
Primary
Human
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.9mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Q9H4F8
SMOC1
Recombinant protein encompassing a sequence within the center region of human SMOC1. The exact sequence is proprietary.
Affinity Purified
RUO
64093
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only