Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMOC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571841
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571841 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571841 Supplier Invitrogen™ Supplier No. PA571841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMOC1 Polyclonal Antibody for Western Blot

Secreted modular calcium-binding protein 1 (SMOC1) is a gene located on chromosome 14q24.2, encoding a member of the SPARC (Secreted Protein, Acidic, and Rich in Cysteine) family of matricellular proteins. SMOC1 is characterized by its multiple domains that facilitate calcium binding and interaction with various components of the extracellular matrix. This protein plays a crucial role in modulating cell-matrix interactions and signaling pathways involved in cellular adhesion, migration, and differentiation. SMOC1 is expressed in multiple tissues, including the eyes, limbs, and the central nervous system, and is essential for developmental processes such as angiogenesis and tissue formation. Mutations in SMOC1 have been linked to ocular conditions like anterior segment dysgenesis and microphthalmia, reflecting its significance in eye development. Current studies focus on the protein's broader implications in developmental disorders and its potential as a therapeutic target in diseases associated with extracellular matrix dysregulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMOC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SMOC1
Gene Accession No. Q8BLY1, Q9H4F8
Gene Alias 2600002F22Rik; AI848508; OAS; secreted modular calcium binding protein 1; secreted modular calcium-binding protein 1; SMOC1; SMOC-1; SPARC related modular calcium binding 1; SPARC-related gene protein; SPARC-related modular calcium binding protein 1; SPARC-related modular calcium-binding protein 1; SPARC-related protein; Srg
Gene Symbols SMOC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 284-313 amino acids from the C-terminal region of human SMOC1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64075, 64093
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.