Learn More
Invitrogen™ SMOC2 Monoclonal Antibody (667713)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | SMOC2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 667713 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | SMOC2 |
| Gene Accession No. | Q9H3U7 |
| Gene Alias | bA270C4A.1; bA37D8.1; dJ421D16.1; DTDP1; MST117; MSTP117; MSTP140; secreted modular calcium-binding protein 2; SMAP2; SMAP-2; SMOC2; SMOC-2; smooth muscle associated protein 2; smooth muscle-associated protein 2; SPARC related modular calcium binding 2; SPARC-related modular calcium-binding protein 2; thyroglobulin type-1 repeat containing protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.