Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smooth Muscle Actic Specific Rabbit anti-Human, Unconjugated, Clone: 5H7, Proteintech
SDP

Catalog No. 502256565 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-6565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-6565 Supplier Proteintech Group Inc Supplier No. 800081RR100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Antibody

The smooth muscle actin specific antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human smooth muscle actin specific. This antibody recognizes human, mouse, rat, chicken, pig antigen. The smooth muscle actin specific antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.

Specifications

Antigen Smooth Muscle Actic Specific
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 5H7
Concentration 260 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 59
Target Species Human, Mouse, Rat, Chicken, Pig
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.