Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smoothelin Antibody (885424), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8278M
Description
Smoothelin Monoclonal antibody specifically detects Smoothelin in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Smoothelin | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6525 | |
Store at 4°C in the dark. | |
IgG1 |
Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
885424 | |
Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
FLJ35365, FLJ38597, smoothelin, SMSMO | |
E. coli-derived recombinant human Smoothelin B, Met1-Leu132, Accession # P53814 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction