Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smoothelin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP23793125UL
Description
Smoothelin Polyclonal specifically detects Smoothelin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Smoothelin | |
Polyclonal | |
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:5000 - 1:10000, Immunocytochemistry/Immunofluorescence Reported in (PMD: 29622561)., Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
P53814 | |
SMTN | |
This antibody was developed against a recombinant protein corresponding to amino acids: LAGLDEGALRKLLEVTADLAERRRIRSAIRELQRQELEREEEALASKRFRAERQDNKENWLHSQQREAEQRAALARLAGQLESMNDVEELTALLR | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ35365, FLJ38597, smoothelin, SMSMO | |
Rabbit | |
Affinity Purified | |
RUO | |
6525 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction