Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smoothened Antibody (1015418) [DyLight 405], Novus Biologicals™
SDP

Catalog No. FAB10276E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10276E 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB10276E Supplier Novus Biologicals Supplier No. FAB10276E
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Smoothened Monoclonal antibody specifically detects Smoothened in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen Smoothened
Applications Immunohistochemistry
Classification Monoclonal
Clone 1015418
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias Gx, Protein Gx, SMOHseven transmembrane helix receptor, smoothened (Drosophila) homolog, smoothened homolog, smoothened homolog (Drosophila)
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human Smoothened, Met1-Tyr233, Accession # Q99835
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6608
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.