Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMOX Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SMOX |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SMOX Polyclonal specifically detects SMOX in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SMOX | |
Polyclonal | |
Rabbit | |
Human | |
54498 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:HDKPVNAESQNSVGVFTREEVRNRIRNDPDDPEATKRLKLAMIQQYLKVESCESSSHSMDEVSLSAFGEWTEIPGAHHIIPSGFMRVVE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
C20orf16, chromosome 20 open reading frame 16, EC 1.5.3.16, flavin containing amine oxidase, flavin-containing spermine oxidase, FLJ20746, MGC1010, PAO, PAO-1, PAOh1, Polyamine oxidase 1, putative cyclin G1 interacting protein, SMOdJ779E11.1, spermine oxidase | |
SMOX | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title