Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMOX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561432 Supplier Invitrogen™ Supplier No. PA561432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGFTDVTVLE ASSHIGGRVQ SVKLGHATFE LGATWIHGSH GNPIYHLAEA NGLLEETTDG ERSVGRISLY SKNGVACYLT NHGRRIPKDV VEEFSDLYN Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 98%.

Polyamines are ubiquitous polycationic alkylamines which include spermine, spermidine, putrescine, and agmatine. These molecules participate in a broad range of cellular functions which include cell cycle modulation, scavenging reactive oxygen species, and the control of gene expression. These molecules also play important roles in neurotransmission through their regulation of cell-surface receptor activity, involvement in intracellular signalling pathways, and their putative roles as neurotransmitters. This gene encodes an FAD-containing enzyme that catalyzes the oxidation of spermine to spermadine and secondarily produces hydrogen peroxide. Multiple transcript variants encoding different isoenzymes have been identified for this gene, some of which have failed to demonstrate significant oxidase activity on natural polyamine substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMOX
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMOX
Gene Accession No. Q9NWM0
Gene Alias B130066H01Rik; C20orf16; flavin containing amine oxidase; flavin-containing spermine oxidase; PAO; PAO1; PAO-1; PAOH; PAOh1; Polyamine oxidase 1; putative cyclin G1 interacting protein; SMO; Smox; Spermine oxidase; UNQ3039/PRO9854
Gene Symbols SMOX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMOX. Recombinant protein control fragment (Product #RP-99298).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54498
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.