Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMPD1 Antibody (09), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306597JF549
Description
SMPD1 Monoclonal antibody specifically detects SMPD1 in Human samples. It is validated for ELISASpecifications
SMPD1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
09 | |
ELISA | |
Acid sphingomyelinase, ASMASE, ASMsphingomyelin phosphodiesterase, EC 3.1.4.12, NPD, sphingomyelin phosphodiesterase 1, acid lysosomal | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SMPD1 (BAD93012.1; Met1-Pro628). | |
0.1 mL | |
Neuroscience | |
6609 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction