Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMPD1 Antibody (OTI3H7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274240B
Description
SMPD1 Monoclonal specifically detects SMPD1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SMPD1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SMPD1 | |
Recombinant protein fragment corresponding to amino acids 62-341 of human SMPD1 (NP_000534) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3H7 | |
Western Blot, Immunohistochemistry | |
Acid sphingomyelinase, ASMASE, ASMsphingomyelin phosphodiesterase, EC 3.1.4.12, NPD, sphingomyelin phosphodiesterase 1, acid lysosomal | |
Mouse | |
Affinity Purified | |
Neuroscience | |
6609.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction