Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMPD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102374
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102374 Supplier Invitrogen™ Supplier No. PA5102374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SMPD2.

Converts sphingomyelin to ceramide. Hydrolyze 1-acyl-2-lyso-sn-glycero-3-phosphocholine (lyso-PC) and 1-O-alkyl-2-lyso-sn-glycero-3-phosphocholine (lyso-platelet-activating factor). The physiological substrate seems to be Lyso-PAF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Smpd2
Gene Accession No. O60906, O70572, Q9ET64
Gene Alias AW108287; ISC1; Lyso-PAF-PLC; lyso-platelet-activating factor-phospholipase C; neutral sphingomyelinase; neutral sphingomyelinase 1; NSMA; NSMASE; N-SMase; nSMase1; SMPD2; Sphingomyelin phosphodiesterase 2; sphingomyelin phosphodiesterase 2, neutral; sphingomyelin phosphodiesterase 2, neutral membrane; sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase)
Gene Symbols Smpd2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SMPD2(Accession O60906), corresponding to amino acid residues E158-K208.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20598, 6610, 83537
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.