Learn More
Invitrogen™ SMPD2 Polyclonal Antibody

Description
SMPD2 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | SMPD2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Smpd2 |
| Gene Accession No. | O60906, O70572 |
| Gene Alias | AW108287; ISC1; Lyso-PAF-PLC; lyso-platelet-activating factor-phospholipase C; neutral sphingomyelinase; neutral sphingomyelinase 1; NSMA; NSMASE; N-SMase; nSMase1; SMPD2; Sphingomyelin phosphodiesterase 2; sphingomyelin phosphodiesterase 2, neutral; sphingomyelin phosphodiesterase 2, neutral membrane; sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase) |
| Gene Symbols | Smpd2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.