Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMPD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553732
Description
Immunogen sequence: QFIHHTSKKA DVVLLCGDLN MHPEDLGCCL LKEWTGLHDA YLETRDFKGS EEGNTMVPKN CYVSQQELKP FPFGVRIDYV LYKAVSGFYI SCKSFETTTG FDPHRGTPLS DHEALMATLF VRHSPPQQNP SST Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 82%.
Converts sphingomyelin to ceramide. Hydrolyze 1-acyl-2-lyso-sn-glycero-3-phosphocholine (lyso-PC) and 1-O-alkyl-2-lyso-sn-glycero-3-phosphocholine (lyso-platelet-activating factor). The physiological substrate seems to be Lyso-PAF.
Specifications
SMPD2 | |
Polyclonal | |
Unconjugated | |
Smpd2 | |
AW108287; ISC1; Lyso-PAF-PLC; lyso-platelet-activating factor-phospholipase C; neutral sphingomyelinase; neutral sphingomyelinase 1; NSMA; NSMASE; N-SMase; nSMase1; SMPD2; Sphingomyelin phosphodiesterase 2; sphingomyelin phosphodiesterase 2, neutral; sphingomyelin phosphodiesterase 2, neutral membrane; sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6610 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60906 | |
Smpd2 | |
Recombinant protein corresponding to Human SMPD2. Recombinant protein control fragment (Product #RP-91103). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction