Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SMPD3/NSMase2 Antibody, Novus Biologicals™
SDP

Catalog No. NB462843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB462843 25 μg
NB462844 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB462843 Supplier Novus Biologicals™ Supplier No. NBP33093225UG

Rabbit Polyclonal Antibody

SMPD3/NSMase2 Polyclonal antibody specifically detects SMPD3/NSMase2 in Human samples. It is validated for Western Blot

Specifications

Antigen SMPD3/NSMase2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot : 1:400
Formulation Phosphate buffered saline
Gene Alias EC 3.1.4.12, FLJ22593, MGC138443, Neutral sphingomyelinase 2, Neutral sphingomyelinase II, nSMase2, nSMase-2, sphingomyelin phosphodiesterase 3, sphingomyelin phosphodiesterase 3, neutral membrane (neutral sphingomyelinaseII)
Host Species Rabbit
Immunogen Synthetic peptide derived from human SMPD3/NSMase2
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55512
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.