Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576574 Supplier Invitrogen™ Supplier No. PA576574
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Catalyzes the hydrolysis of sphingomyelin to form ceramide and phosphocholine. Ceramide mediates numerous cellular functions, such as apoptosis and growth arrest, and is capable of regulating these 2 cellular events independently. Also hydrolyzes sphingosylphosphocholine. Regulates the cell cycle by acting as a growth suppressor in confluent cells. Probably acts as a regulator of postnatal development and participates in bone and dentin mineralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPD3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SMPD3
Gene Accession No. O35049, Q9JJY3, Q9NY59
Gene Alias 4631433G07Rik; AI427456; AW537966; Cca1; confluent 3Y1 cell-associated 1; confluent 3Y1 cell-associated protein 1; fro; neutral sphingomyelin phosphodiesterase 3; neutral sphingomyelinase 2; Neutral sphingomyelinase II; NSMASE2; nSMase-2; SMPD3; Sphingomyelin phosphodiesterase 3; sphingomyelin phosphodiesterase 3, neutral; sphingomyelin phosphodiesterase 3, neutral membrane; sphingomyelin phosphodiesterase 3, neutral membrane (neutral sphingomyelinase II)
Gene Symbols SMPD3
Host Species Rabbit
Immunogen Recombinant fusion protein of human SMPD3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55512, 58994, 94338
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.