Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMPD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110401
Description
Immunogen sequence: ITQAKHTAKS ISDQCAESPA GHSFLSWLGF SSMDTNGSYT ANDLDEMGQD SVRKTDEYLE KALEYLRQIF RLSEAQLRQF TLALGTTQDE NGKKQLPDCI VGEDGLILTP LGRYQIINGL RRFEIEYQGD PELQPIRSYE IASLVRTLFR LSSAINHRFA GQMAALCSRD DFLGSFCRYH LTEPGLASRH LLSPVGRRQV A.
The protein encoded by SMPD4 is a sphingomyelinase that catalyzes the hydrolysis of membrane sphingomyelin to form phosphorylcholine and ceramide. This gene is activated by DNA damage, cellular stress, and tumor necrosis factor, but it is downregulated by wild-type p53. The encoded protein localizes to the endoplasmic reticulum and Golgi network.
Specifications
| SMPD4 | |
| Polyclonal | |
| Unconjugated | |
| SMPD4 | |
| 4122402O22Rik; Kiaa1418; mKIAA1418; NET13; neutral membrane (neutral sphingomyelinase-3); neutral sphingomyelinase 3; Neutral sphingomyelinase III; neutral sphingomyelinase-3; NSMASE3; nSMase-3; RGD1310674; SKNY; SKNY protein; Smpd4; Sphingomyelin phosphodiesterase 4; sphingomyelin phosphodiesterase 4, neutral membrane; sphingomyelin phosphodiesterase 4, neutral membrane (neutral sphingomyelinase-3) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55627, 77626 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.08 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6ZPR5, Q9NXE4 | |
| SMPD4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 600-800 of human SMPD4 (NP_060421.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction