Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMPDL3A Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP320170C
Description
SMPDL3A Polyclonal antibody specifically detects SMPDL3A in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SMPDL3A | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
0610010C24Rik, acid sphingomyelinase-like phosphodiesterase 3a, ASM3A, ASML3a, ASM-like phosphodiesterase 3a, EC 3.1.4, EC 3.1.4.-, FLJ20177, sphingomyelin phosphodiesterase, acid-like 3A, yR36GH4.1 | |
Produced in rabbits immunized with purified, recombinant human SMPDL3A (Uniprot number: Q92484; Met1-Tyr453) | |
0.1 mL | |
Lipid and Metabolism | |
10924 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction