Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMPDL3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556367 Supplier Invitrogen™ Supplier No. PA556367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPVNSLFVAP AVTPVKSVLE KQTNNPGIRL FQYDPRDYKL LDMLQYYLNL TEANLKGESI WKLEYILTQT YDIEDLQPES LYGLAKQFTI LDSKQFIKYY NYFFVSYDSS ATCDKTCKAF QICAIM Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%.

Has in vitro nucleotide phosphodiesterase activity with nucleoside triphosphates, such as ATP (PubMed:25288789, PubMed:26783088). Has in vitro activity with p-nitrophenyl-TMP (PubMed:25288789). Has lower activity with nucleoside diphosphates, and no activity with nucleoside monophosphates (PubMed:25288789, PubMed:26783088). Has in vitro activity with CDP-choline, giving rise to CMP and phosphocholine. Has in vitro activity with CDP-ethanolamine (PubMed:26783088). Does not have sphingomyelin phosphodiesterase activity (PubMed:25288789, PubMed:26783088). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPDL3A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMPDL3A
Gene Accession No. Q92484
Gene Alias 0610010C24Rik; Acid sphingomyelinase-like phosphodiesterase 3a; AI529588; ASM3A; ASML3; ASML3A; ASM-like phosphodiesterase 3a; hypothetical LOC541525; si:ch211-199i15.3; SMPDL3A; sphingomyelin phosphodiesterase acid like 3A; sphingomyelin phosphodiesterase acid-like 3A; sphingomyelin phosphodiesterase, acid-like 3A; yR36GH4.1; zgc:113352
Gene Symbols SMPDL3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMPDL3A. Recombinant protein control fragment (Product #RP-101899).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10924
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.