Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMPDL3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540798
Description
Peptide sequence: ILWTGDDTPH VPDEKLGEAA VLEIVERLTK LIREVFPDTK VYAALGNHDF Sequence homology: Cow: 85%; Dog: 85%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 85%; Rabbit: 92%; Rat: 85%; Zebrafish: 85%.
Located on chromosome 1, this gene encodes for acid sphingomyelinase-like phosphodiesterase 3b precursor protein.
Specifications
| SMPDL3B | |
| Polyclonal | |
| Unconjugated | |
| SMPDL3B | |
| 1110054A24Rik; Acid sphingomyelinase-like phosphodiesterase 3b; Asml3b; ASM-like phosphodiesterase 3b; ASMLPD; AU045240; im:7137990; si:ch1073-55d10.1; Smpdl3b; sphingomyelin phosphodiesterase acid like 3B; sphingomyelin phosphodiesterase acid-like 3B; sphingomyelin phosphodiesterase, acid-like 3B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27293 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q92485 | |
| SMPDL3B | |
| Synthetic peptide directed towards the N-terminal of human SMPDL3B (aa 88-137). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction