Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMR3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569168
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569168 Supplier Invitrogen™ Supplier No. PA569168
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 75%; Horse: 91%; Human: 100%; Rat: 86%.

SMR3A may play a role in protection or detoxification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMR3A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SMR3A
Gene Accession No. Q99954
Gene Alias P-B1; PBI; PRL5; PROL5; proline rich 5 (salivary); proline- rich protein 5; proline-rich protein 5; Proline-rich protein PBI; protein homologous to salivary proline-rich protein P-B; PR-Vbeta1; SMR3A; submaxillary gland androgen regulated protein 3 homolog A; submaxillary gland androgen regulated protein 3A; submaxillary gland androgen-regulated protein 3A; variable coding sequence B1; Vcsb1; VCS-beta1
Gene Symbols SMR3A
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SMR3A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26952
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.