Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMRT / NCoR2 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992902
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2902 10 μL
50-199-2901 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2902 Supplier Active Motif Supplier No. 39846
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Steroid and thyroid hormones and retinoic acid regulate a complex array of gene expression activity via intracellular receptor transcription factors belonging to the ligand-dependent nuclear receptor superfamily. Adding to the complexity of function of these transcription factors are associated proteins known as coactivators and corepressors which, as their names suggest, enhance or depress transcription activity of the nuclear receptor with which they associate. Silencing mediator of retinoic acid and thyroid hormone receptor (SMRT) and nuclear receptor corepressor (N-CoR) are related transcriptional corepressors which contain two distinct domains capable of interacting with unliganded nuclear receptors to repress their basal transcriptional activity.

Specifications

Antigen SMRT / NCoR2
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene NCOR2
Gene Accession No. Q9Y618
Gene Alias CTG repeat protein 26; CTG26; N-CoR; NCOR2; N-CoR2; Nuclear receptor corepressor 2; nuclear receptor co-repressor 2; silencing mediator for retinoid and thyroid hormone receptors; Silencing mediator of retinoic acid and thyroid hormone receptor; silencing mediator of retinoid acid and thyroid hormone receptor; SMAP270; Smrt; SMRTe; SMRTE-tau; T3 receptor-associating factor; Thyroid-, retinoic-acid-receptor-associated corepressor; TNRC14; TRAC; TRAC1; TRAC-1
Gene Symbols NCOR2
Host Species Rabbit
Immunogen This SMRT / NCoR2 antibody was raised against a peptide within the N-terminal region of SMRT / NCoR2.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9612
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.