Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583108
Description
Immunogen sequence: GDGGILCEIV KLKPKMVTMV EIDQMVIDGC KKYMRKTCGD VLDNLKGDCY QVLIEDCIPV LKRYAKEGRE FDYVINDLTA VPISTSPE.
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
Specifications
SMS | |
Polyclonal | |
Unconjugated | |
SMS | |
AI427066; Gy; gyro; MRSR; SMS; Spermidine aminopropyltransferase; spermine synthase; SpmST; SPMSY; SpS; SRS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6611 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P52788 | |
SMS | |
Recombinant protein corresponding to Human SMS. Recombinant protein control fragment (Product #RP-95185). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction