Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583109
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583109 Supplier Invitrogen™ Supplier No. PA583109
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSTLDFMLGA KADGETILKG LQSIFQEQGM AESVHTWQDH GYLATYTNKN GSFANLRIYP HGLVLLDLQS YDGDAQGKEE IDSILNKVEE RMKELS Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.

Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMS
Gene Accession No. P52788
Gene Alias AI427066; Gy; gyro; MRSR; SMS; Spermidine aminopropyltransferase; spermine synthase; SpmST; SPMSY; SpS; SRS
Gene Symbols SMS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMS. Recombinant protein control fragment (Product #RP-95184).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6611
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.