Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571762
Description
SMT3 Polyclonal Antibody for ELISA
Covalent modification of target lysines by SUMO (small ubiquitin-like modifier) modulates processes such as protein localization, transcription, nuclear transport, mitosis, DNA replication and repair, signal transduction, and viral reproduction. SUMO does not seem to be involved in protein degradation and may in fact function as an antagonist of ubiquitin in the degradation process. In the development of Drosophila, SUMO plays a maternal role in anterior-posterior (A/P) polarity and patterning.
Specifications
SMT3 | |
Polyclonal | |
Unconjugated | |
smt3 | |
anon-EST:Posey240; CG4494; CG4494-PA; CG4494-PB; D9719.15; Dm0342; Dmel\CG4494; Dmel_CG4494; Dmsmt3; DmSUMO-1; dSmt3; l(2)04493; l(2)SH0182; l(2)SH2 0182; LD07775p; lethal (2) SH0182; small Ubiquitin-like modifier; smt3; smt3-PA; smt3-PB; SUMO; SUMO family protein SMT3; SUMO protein; ubiquitin-like protein SMT3; YDR510W | |
Rabbit | |
Ammonium Sulfate Precipitation, Size-exclusion - Dialysis | |
RUO | |
33981 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
smt3 | |
KLH conjugated synthetic peptide selected from the N-terminal region of Drosophila SUMO1. | |
400 μL | |
Primary | |
Human, Drosophila | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction