Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMTNL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548211
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548211 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548211 Supplier Invitrogen™ Supplier No. PA548211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMTNL1 Polyclonal Antibody for Western Blot, Flow

Plays a role in the regulation of contractile properties of both striated and smooth muscles. When unphosphorylated, may inhibit myosin dephosphorylation. Phosphorylation at Ser-299 reduces this inhibitory activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMTNL1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SMTNL1
Gene Accession No. A8MU46
Gene Alias 1110030K22Rik; calponin homology-associated smooth muscle protein; CHASM; smoothelin like 1; smoothelin-like 1; smoothelin-like protein 1; Smtnl1
Gene Symbols SMTNL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 130-159 amino acids from the Central region of human SMTNL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219537
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.