Learn More
SMUG1 Mouse anti-Human, Clone: OTI5G1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810345

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SMUG1 is a glycosylase that removes uracil from single- and double-stranded DNA in nuclear chromatin, thus contributing to base excision repair.Specifications
SMUG1 | |
Monoclonal | |
Unconjugated | |
SMUG1 | |
FDG, HMUDG, UNG3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23583 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5G1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q53HV7 | |
SMUG1 | |
Full length human recombit protein of human SMUG1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.