Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMUG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575522
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
SMUG1 is a glycosylase that removes uracil from single- and double-stranded DNA in nuclear chromatin, thus contributing to base excision repair.
Specifications
SMUG1 | |
Polyclonal | |
Unconjugated | |
SMUG1 | |
1200013B09Rik; A930006H09Rik; C85220; FDG; HMUDG; MGC104370; single-strand selective monofunctional uracil DNA glycosylase; Single-strand selective monofunctional uracil-DNA glycosylase; single-strand-selective monofunctional uracil-DNA glycosylase 1; SMUG1; UNG3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23583, 315344, 71726 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53HV7, Q6P5C5, Q811Q1 | |
SMUG1 | |
Synthetic peptide corresponding to amino acids 137-182 of Human SMUG1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction