Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMUG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120070 Supplier Invitrogen™ Supplier No. PA5120070
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: LO2, Mouse kidney, Mouse liver, Mouse lung. The target is usually found in the following locations: Nucleus. Immunogen sequence: MPQAFLLGSI HEPAGALMEP QPCPGSLAES FLEEELRLNA ELSQLQFSEP VGIIYNPVEY AWEPHRNYVT RYCQGPKEVL FLGMNPGPFG MAQTGVPFGE VSMVRDWLGI VGPVLTPPQE HPKRPVLGLE CPQSEVSGAR FWGFFRNLCG QPEVFFHHCF VHNLCPLLFL APSGRNLTPA ELPAKQREQL LGICDAALCR QVQLLGVRLV VGVGRLAEQR ARRALAGLMP EVQVEGLLHP SPRNPQANKG WEAVAKERLN ELGLLPLLLK.

SMUG1 is a glycosylase that removes uracil from single- and double-stranded DNA in nuclear chromatin, thus contributing to base excision repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMUG1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SMUG1
Gene Accession No. Q53HV7, Q6P5C5
Gene Alias 1200013B09Rik; A930006H09Rik; C85220; FDG; HMUDG; MGC104370; single-strand selective monofunctional uracil DNA glycosylase; Single-strand selective monofunctional uracil-DNA glycosylase; single-strand-selective monofunctional uracil-DNA glycosylase 1; SMUG1; UNG3
Gene Symbols SMUG1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human SMUG1 (NP_055126.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23583, 71726
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.