Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMUG1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE1798MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1798MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1798MI Supplier MilliporeSigma Supplier No. ABE1798
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-SMUG1 is a highly specific rabbit polyclonal antibody that targets single-strand selective monofunctional uracil DNA glycosylase and has been tested in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMUG1
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols SMUG1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 13 amino acids from the N-terminal region of human SMUG1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_055126
Test Specificity This polyclonal antibody detects SMUG1 in HeLa cell nuclear extract. It targets an epitope within 13 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.