Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMURF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA511943
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA511943 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA511943 Supplier Invitrogen™ Supplier No. PA511943
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with Xenopus and zebrafish based on sequence homology.

Members of the transforming growth factor-beta (TGFB) family signal through type I and type II serine/threonine-kinase receptors, which in turn activate the SMAD signaling pathway. Bone morphogenetic protein (BMP) receptors target SMAD1, SMAD5, and SMAD8, whereas receptors for activin and TGFB (e. g., ACVR1 and TGFBR1, respectively) target SMAD2 and SMAD3. Phosphorylation of these receptor-regulated SMADs induces their association with the common-partner SMAD, SMAD4. Smurf1, a HECT domain E3 ubiquitin ligase, regulates cell polarity and protrusive activity and is required to maintain the transformed morphology and motility of a tumor cell. Atypical protein kinase C-zeta (PKC2), an effector of the Cdc42/Rac1-PAR6 polarity complex, recruits Smurf1 to cellular protrusions, where it controlled the local level of RhoA. Smurf1 thus links the polarity complex to degradation of RhoA in lamellipodia and filopodia to prevent RhoA signaling during dynamic membrane movements.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMURF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Smurf1
Gene Accession No. Q9CUN6, Q9HCE7
Gene Alias 4930431E10Rik; E3 ubiquitin ligase SMURF1; E3 ubiquitin ligase SMURF2; E3 ubiquitin-protein ligase SMURF1; E3 ubiquitin-protein ligase SMURF2; HECT-type E3 ubiquitin transferase SMURF1; HECT-type E3 ubiquitin transferase SMURF2; hSMURF1; hSMURF2; KIAA1625; mKIAA1625; RGD1309707; SMAD specific E3 ubiquitin protein ligase 1; SMAD specific E3 ubiquitin protein ligase 2; Smad ubiquitination regulatory factor 1; SMAD ubiquitination regulatory factor 2; Smad-specific E3 ubiquitin ligase 1; SMAD-specific E3 ubiquitin-protein ligase 1; SMAD-specific E3 ubiquitin-protein ligase 2; SMURF1; SMURF2
Gene Symbols Smurf1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 66-96 amino acids from the N-terminal region of human SMURF1.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57154, 75788
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.