Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMURF2 Antibody (OTI3E4), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274241JF646
Description
SMURF2 Monoclonal specifically detects SMURF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SMURF2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SMURF2 | |
Recombinant protein fragment corresponding to amino acids 151-440 of human SMURF2 (NP_073576) produced in E.coli. | |
0.1 mL | |
64750.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3E4 | |
Western Blot, Immunohistochemistry | |
DKFZp686F0270, E3 ubiquitin ligase SMURF2, E3 ubiquitin-protein ligase SMURF2, EC 6.3.2, EC 6.3.2.-, hSMURF2, MGC138150, SMAD specific E3 ubiquitin protein ligase 2, SMAD ubiquitination regulatory factor 2, SMAD-specific E3 ubiquitin-protein ligase 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction