Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMURF2 Rabbit anti-Human, Clone: JU32-34, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275667
Description
SMURF2 Monoclonal specifically detects SMURF2 in Human samples. It is validated for Western Blot.Specifications
SMURF2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 | |
DKFZp686F0270, E3 ubiquitin ligase SMURF2, E3 ubiquitin-protein ligase SMURF2, EC 6.3.2, EC 6.3.2.-, hSMURF2, MGC138150, SMAD specific E3 ubiquitin protein ligase 2, SMAD ubiquitination regulatory factor 2, SMAD-specific E3 ubiquitin-protein ligase 2 | |
Rabbit | |
86 kDa | |
100 μL | |
64750.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
JU32-34 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
SMURF2 | |
Recombinant protein within Human SMURF2 aa 150-340. (SwissProt: Q9HAU4 Human) | |
Protein A purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction