Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMYD1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989758
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9758 Supplier Bioss Supplier No. BS12647R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acts as a transcriptional repressor. Essential for cardiomyocyte differentiation and cardiac morphogenesis.

Specifications

Antigen SMYD1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene Smyd1
Gene Accession No. Q8NB12
Gene Alias 4632404M21Rik; Bop; C78565; CD8 beta opposite; CD8beta opposite strand; CD8b-opposite; Histone-lysine N-methyltransferase Smyd1; KMT3D; m-BOP; SET and MYND domain containing 1; SET and MYND domain-containing protein 1; Smyd1; Zinc finger protein BOP; zinc finger, MYND domain containing 18; ZMYND18; ZMYND22
Gene Symbols Smyd1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SMYD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150572
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.