Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMYD2 Monoclonal Antibody (8E5)
GREENER_CHOICE

Catalog No. PIMA532965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532965 Supplier Invitrogen™ Supplier No. MA532965
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Purity is > 95% by SDS-PAGE.

SET domain-containing proteins, such as SMYD2, catalyze lysine methylation..
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMYD2
Applications Western Blot
Classification Monoclonal
Clone 8E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SMYD2
Gene Accession No. Q8R5A0, Q9NRG4
Gene Alias 1110020E07Rik; 4930402C15; Histone methyltransferase SMYD2; HSKM-B; KMT3C; lysine N-methyltransferase 3C; N-lysine methyltransferase SMYD2; SET and MYND domain containing 2; SET and MYND domain-containing protein 2; Smyd2; zinc finger, MYND domain containing 14; ZMYND14
Gene Symbols SMYD2
Host Species Mouse
Immunogen Recombinant full length Human SMYD2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226830, 56950
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.