Learn More
Invitrogen™ SMYD2 Polyclonal Antibody

Description
SMYD2 Polyclonal Antibody detects endogenous levels of SMYD2 protein.
Specifications
Specifications
| Antigen | SMYD2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | SMYD2 |
| Gene Accession No. | Q7M6Z3, Q8R5A0, Q9NRG4 |
| Gene Alias | 1110020E07Rik; 4930402C15; Histone methyltransferase SMYD2; HSKM-B; KMT3C; lysine N-methyltransferase 3C; N-lysine methyltransferase SMYD2; SET and MYND domain containing 2; SET and MYND domain-containing protein 2; Smyd2; zinc finger, MYND domain containing 14; ZMYND14 |
| Gene Symbols | SMYD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.