Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SMYD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115938
Description
Antibody detects endogenous levels of total SMYD2.
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation..
Specifications
SMYD2 | |
Polyclonal | |
Unconjugated | |
SMYD2 | |
1110020E07Rik; 4930402C15; Histone methyltransferase SMYD2; HSKM-B; KMT3C; lysine N-methyltransferase 3C; N-lysine methyltransferase SMYD2; SET and MYND domain containing 2; SET and MYND domain-containing protein 2; Smyd2; zinc finger, MYND domain containing 14; ZMYND14 | |
Rabbit | |
Affinity chromatography | |
RUO | |
226830, 289372, 56950 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q7M6Z3, Q8R5A0, Q9NRG4 | |
SMYD2 | |
A synthesized peptide derived from human SMYD2(Accession Q9NRG4), corresponding to amino acid residues Y258-A308. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction