Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMYD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SMYD3 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SMYD3 Polyclonal specifically detects SMYD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SMYD3 | |
Polyclonal | |
Rabbit | |
Human | |
bA74P14.1, bA74P14.1 (novel protein), EC 2.1.1, EC 2.1.1.43, FLJ21080, KMT3E, MGC104324, MYND domain containing 1, SET and MYND domain containing 3, SET and MYND domain-containing protein 3, Zinc finger MYND domain-containing protein 1, zinc finger protein, subfamily 3A (MYND domain containing), 1, ZMYND1, ZNFN3A1 | |
SMYD3 | |
IgG | |
Affinity Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
64754 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GELLFRSDPLAYTVCKGSRGVVCDRCLLGKEKLMRCSQCRVAKYCSAKCQKKAWPDHKRECKCLKSCKPRYPPDSVR | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title