Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMYD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584195 Supplier Invitrogen™ Supplier No. PA584195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GELLFRSDPL AYTVCKGSRG VVCDRCLLGK EKLMRCSQCR VAKYCSAKCQ KKAWPDHKRE CKCLKSCKPR YPPDSVR Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 35%.

SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMYD3
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMYD3
Gene Accession No. Q9H7B4
Gene Alias 2410008A19Rik; bA74P14.1; bA74P14.1 (novel protein); histone-lysine N-methyltransferase SMYD3; KMT3E; SET and MYND domain containing 3; SET and MYND domain-containing protein 3; Smyd3; Zinc finger MYND domain-containing protein 1; zinc finger protein, subfamily 3A (MYND domain containing), 1; zinc finger, MYND domain containing 1; ZMYND1; ZNFN3A1
Gene Symbols SMYD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMYD3. Recombinant protein control fragment (Product #RP-106533).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64754
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.