Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMYD4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179406
Description
SMYD4 Polyclonal specifically detects SMYD4 in Human samples. It is validated for Western Blot.Specifications
SMYD4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
KIAA1936ZMYND21, SET and MYND domain containing 4, SET and MYND domain-containing protein 4 | |
Rabbit | |
Affinity purified | |
RUO | |
114826 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_443160 | |
SMYD4 | |
Synthetic peptide directed towards the N terminal of human SMYD4The immunogen for this antibody is SMYD4. Peptide sequence EGNKKFQEKDYTGAAVLYSKGVSHSRPNTEDMSLCHANRSAALFHLGQYE. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Rabbit: 92%; Pig: 85%; Canine: 84%. | |
Human, Mouse, Rat, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction