Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMYD5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553314
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553314 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553314 Supplier Invitrogen™ Supplier No. PA553314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLELLRRLFT EALYEEAVSQ WFTPDGFRSL FALVGTNGQG IGTSSLSQWV HACDTLELKP QDREQLDAFI DQLYKDIEAA TGEFLNCEGS GLFVLQSCC Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 95%.

Retinoic acid (RA) represents the oxidized form of vitamin A and, via interactions with retinoic acid receptors (RARs), plays a crucial role in development, cellular growth and differentiation. The gene encoding RAI15 maps to human chromosome 2, which houses over 1,400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes. Additionally, an extremely rare recessive genetic disorder, Alström syndrome, is caused by mutations in the ALMS1 gene, which maps to chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMYD5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMYD5
Gene Accession No. Q6GMV2
Gene Alias AW536703; NN8-4AG; Protein NN8 4AG; Protein NN8-4AG; Rai15; retinoic acid induced 15; retinoic acid responsive; retinoic acid responsive gene 1; retinoic acid-induced protein 15; RRG1; SET and MYND domain containing 5; SET and MYND domain-containing protein 5; SMYD family member 5; SMYD5; ZMYND23
Gene Symbols SMYD5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SMYD5. Recombinant protein control fragment (Product #RP-91632).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10322
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.