Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SN 38
GREENER_CHOICESDP

Catalog No. 268410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-841-0 10 mg
26-845-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-841-0 Supplier Tocris Bioscience™ Supplier No. 2684/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

DNA topoisomerase I inhibitor; antitumor

Active metabolite of CPT-11 (Cat. No. 2688) that inhibits DNA topoisomerase I (IC50 values are 0.74 and 1.9 μM in P388 and Ehrlich cells respectively). Inhibits DNA and RNA synthesis (IC50 values are 0.077 and 1.3 μM respectively) but does not affect protein synthesis. Displays potent antitumor activity against a range of human tumor cell lines (IC50 values are 3.3, 13, 19 and 22 nM for HCT-116, BEL-7402, HL60 and HELA cells respectively).

Chemical Identifiers

CAS 86639-52-3
Molecular Formula C22H20N2O5

Specifications

Quantity 10 mg
Formula Weight 392.4
Purity >98%
Molecular Weight (g/mol) 392.411
Chemical Name or Material SN 38
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.