Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SN-6
GREENER_CHOICE

Catalog No. 218450
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-845-0 50 mg
21-841-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-845-0 Supplier Tocris Bioscience™ Supplier No. 2184/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective Na+/Ca2+ exchange inhibitor (reverse mode)

SN-6 is a selective Na+/Ca2+-exchange (NCX) inhibitor; displays some selectivity for NCX1. IC50 values are 2.9, 16 and 8.6 μM for inhibition of intracellular Na+-dependent 45Ca2+ uptake by cells expressing NCX1, NCX2 and NCX3 respectively. Has some affinity for mACh receptors (IC50 = 18 μM) but minimal activity against NCKX2 and various receptors and ion channels (IC50 > 30 μM). Preferentially blocks Ca2+ influx mode and is more selective for NCX isoforms than KB-R7943 (Cat. No. 1244). Anti-ischemic; potently protects against hypoxia-induced renal tubular cell damage (IC50 = 0.63 μM).

Specifications

Quantity 50 mg
Product Type SN-6
Purity >99%
Molecular Weight (g/mol) 402.16
Inhibitors SN-6

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.