Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SN38 Recombinant Rabbit Monoclonal Antibody (PSH0-05)
GREENER_CHOICE

Catalog No. FEMA550767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550767 Supplier Invitrogen™ Supplier No. MA550767
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Product MA5-50767 was used in a Competitive ELISA and Indirect ELISA.

SN38 is a chemical compound that belongs to the class of topoisomerase I inhibitors. It is the active metabolite of irinotecan, a chemotherapeutic drug used in the treatment of various types of cancer, particularly colorectal cancer. SN38 exerts its anti-cancer effects by inhibiting the action of topoisomerase I, an enzyme involved in DNA replication and repair. By inhibiting this enzyme, SN38 prevents DNA from unwinding, leading to the accumulation of DNA damage and ultimately inducing cell death in cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SN38
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone PSH0-05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias SN-38
Host Species Rabbit
Immunogen SN38-OVA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical, Rabbit
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Rabbit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.