Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAI2 Monoclonal Antibody (OTI1H10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504929S


Description
SNAI2 Monoclonal antibody specifically detects SNAI2 in Human samples. It is validated for Western BlotSpecifications
SNAI2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P20073 | |
Mouse | |
Affinity chromatography | |
RUO | |
310 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI1H10 | |
Unconjugated | |
ANXA7 | |
MGC10182; neural crest transcription factor Slug; Protein snail homolog 2; SLUG; slug (chicken homolog), zinc finger protein; Slug, chicken homolog; Slugh; SLUGH1; Snai2; snail family transcriptional repressor 2; snail family zinc finger 2; snail homolog 2; Snail2; snail-like protein 2; WS2D; Zinc finger protein SNAI2; zinc finger transcription factor snail 2 | |
Human recombinant protein fragment corresponding to amino acids 105-411 of human ANXA7 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction