Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNAI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531970 Supplier Invitrogen™ Supplier No. PA531970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Rhesus Monkey (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SNAI3 is a member of the SNAIL gene family, named for the Drosophila snail gene, which plays roles in mesodermal formation during embryogenesis (Katoh and Katoh, 2003).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAI3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene SNAI3
Gene Accession No. Q3KNW1
Gene Alias AI643946; Protein snail homolog 3; Smuc; SNAI3; snail family transcriptional repressor 3; snail family zinc finger 3; snail homolog 3; SNAIL3; snail-related gene from muscle cells; snail-related protein 2; Zfp293; zinc finger protein 293; Zinc finger protein SNAI3; ZNF293
Gene Symbols SNAI3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SNAI3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333929
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.